missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)pyridine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. T3155100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3155100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3155100MG Supplier TCI America Supplier No. T3155100MG
Only null left

Chemical Identifiers

CAS 87407-12-3
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD07375386
InChI Key DMWLBOPLZYJGPT-UHFFFAOYSA-N
Synonym 3-trifluoromethyl pyridine-2-carboxylic acid, 3-trifluoromethyl picolinic acid, 3-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 3-trifluoromethyl, 3-trifluoromethyl-2-picolinic acid, acmc-209qkb, ksc447o0j, 3-trifluoromethylpicolinic acid, trifluoromethyl-2-picolinic acid
PubChem CID 13233844
IUPAC Name 3-(trifluoromethyl)pyridine-2-carboxylic acid
SMILES OC(=O)C1=C(C=CC=N1)C(F)(F)F

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 100 mg
Formula Weight 191.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethyl)pyridine-2-carboxylic Acid