missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenylacetic Acid 98.0+%, TCI America™

Catalog No. T16625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16625G 5 g
T166225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16625G Supplier TCI America Supplier No. T16625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 351-35-9
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00004339
InChI Key BLXXCCIBGGBDHI-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride
PubChem CID 67695
IUPAC Name 2-[3-(trifluoromethyl)phenyl]acetic acid
SMILES C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 5 g
Formula Weight 204.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethyl)phenylacetic Acid