missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenylacetic acid, 96%

Catalog No. AAL1096018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1096018 50 g
AAL1096009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1096018 Supplier Thermo Scientific Chemicals Supplier No. L1096018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 351-35-9 | C9H7F3O2 | 204.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 351-35-9
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00004339
InChI Key BLXXCCIBGGBDHI-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride
PubChem CID 67695
IUPAC Name 2-[3-(trifluoromethyl)phenyl]acetic acid
SMILES C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O

Specifications

Melting Point 77°C to 78°C
Quantity 50 g
Beilstein 2213223
Formula Weight 204.15
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)phenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-511-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only