missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzyl mercaptan, tech. 90%

Catalog No. AAL0815506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0815506 5 g
AAL0815503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0815506 Supplier Thermo Scientific Chemicals Supplier No. L0815506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25697-55-6 | C8H7F3S | 192.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25697-55-6
Molecular Formula C8H7F3S
Molecular Weight (g/mol) 192.199
MDL Number MFCD00042435
InChI Key CQIQWIMXCPTQPJ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl mercaptan, 3-trifluoromethyl phenyl methanethiol, 3-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl phenylmethanethiol, 3-trifluoromethyl benzylthiol, m-trifluoromethylbenzyl mercaptan, 3-trifluoromethyl benzylmercaptan, 3-trifluoromethyl phenyl methane-1-thiol, acmc-1cnzu
PubChem CID 141236
IUPAC Name [3-(trifluoromethyl)phenyl]methanethiol
SMILES C1=CC(=CC(=C1)C(F)(F)F)CS

Specifications

Boiling Point 75°C to 77°C (5 mmHg)
Refractive Index 1.49
Quantity 5 g
UN Number UN2810
Beilstein 2520097
Sensitivity Air Sensitive
Formula Weight 192.2
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-(Trifluoromethyl)benzyl mercaptan
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only