missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzonitrile, 98%

CAS: 368-77-4 | C8H4F3N | 171.122 g/mol

Supplier:  Thermo Scientific Chemicals A1702922

 View more versions of this product

Catalog No. AAA1702922


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(Trifluoromethyl)benzonitrile
14°C to 18°C
188°C to 190°C
C8H4F3N
MFCD00001807
UN3276
3-trifluoromethyl benzonitrile, 3-cyanobenzotrifluoride, m-trifluoromethylbenzonitrile, alpha,alpha,alpha-trifluoro-m-tolunitrile, benzonitrile, 3-trifluoromethyl, a,a,a-trifluoro-m-tolunitrile, unii-y7zd7dk3ay, alpha,alpha,alpha-trifluoro-m-toluonitrile, y7zd7dk3ay, a,a,a-trifluor-m-tolunitril
C1=CC(=CC(=C1)C(F)(F)F)C#N
171.122
171.12
368-77-4
1.28
72°C (161°F)
1.459
100 g
1868102
OGOBINRVCUWLGN-UHFFFAOYSA-N
3-(trifluoromethyl)benzonitrile
67783
98%

Chemical Identifiers

368-77-4
171.122
OGOBINRVCUWLGN-UHFFFAOYSA-N
67783
C1=CC(=CC(=C1)C(F)(F)F)C#N
C8H4F3N
MFCD00001807
3-trifluoromethyl benzonitrile, 3-cyanobenzotrifluoride, m-trifluoromethylbenzonitrile, alpha,alpha,alpha-trifluoro-m-tolunitrile, benzonitrile, 3-trifluoromethyl, a,a,a-trifluoro-m-tolunitrile, unii-y7zd7dk3ay, alpha,alpha,alpha-trifluoro-m-toluonitrile, y7zd7dk3ay, a,a,a-trifluor-m-tolunitril
3-(trifluoromethyl)benzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 206-711-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only