accessibility menu, dialog, popup

UserName

FFFN

CAS RN 368-77-4

IUPAC Name: 3-(trifluoromethyl)benzonitrile
Synonyms: A,A,A-trifluor-M-tolunitril
Molecular Weight (g/mol): 171.12
Molecular Formula: C8H4F3N
InChi Key: OGOBINRVCUWLGN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)C#N
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)