missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzonitrile, 98%

Catalog No. p-7023459
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1702906 5 g
AAA1702914 25 g
AAA1702922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1702906 Supplier Thermo Scientific Chemicals Supplier No. A1702906
Only null left

CAS: 368-77-4 | C8H4F3N | 171.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 368-77-4
Molecular Formula C8H4F3N
Molecular Weight (g/mol) 171.122
MDL Number MFCD00001807
InChI Key OGOBINRVCUWLGN-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzonitrile, 3-cyanobenzotrifluoride, m-trifluoromethylbenzonitrile, alpha,alpha,alpha-trifluoro-m-tolunitrile, benzonitrile, 3-trifluoromethyl, a,a,a-trifluoro-m-tolunitrile, unii-y7zd7dk3ay, alpha,alpha,alpha-trifluoro-m-toluonitrile, y7zd7dk3ay, a,a,a-trifluor-m-tolunitril
PubChem CID 67783
IUPAC Name 3-(trifluoromethyl)benzonitrile
SMILES C1=CC(=CC(=C1)C(F)(F)F)C#N

Specifications

Melting Point 14°C to 18°C
Density 1.28
Boiling Point 188°C to 190°C
Flash Point 72°C (161°F)
Refractive Index 1.459
Quantity 5 g
UN Number UN3276
Beilstein 1868102
Formula Weight 171.12
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzonitrile
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 206-711-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only