missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethyl)benzenethiol 95.0+%, TCI America™
Supplier: TCI America T278025G
Specifications
| 3-(Trifluoromethyl)benzenethiol | |
| Yellow | |
| C7H5F3S | |
| 25 g | |
| 3-trifluoromethyl thiophenol, 3-trifluoromethyl benzenethiol, 3-trifluoromethyl benzene-1-thiol, 3-trifluoromethylthiophenol, m-trifluoromethylthiophenol, 3-mercaptobenzotrifluoride, m-trifluoromethyl benzenethiol, benzenethiol, 3-trifluoromethyl, pubchem4600 | |
| FC(F)(F)C1=CC=CC(S)=C1 | |
| 178.17 | |
| 178.17 | |
| Liquid |
| 937-00-8 | |
| 163°C | |
| MFCD00041142 | |
| 2810 | |
| SCURCOWZQJIUGR-UHFFFAOYSA-N | |
| 3-(trifluoromethyl)benzene-1-thiol | |
| 136751 | |
| ≥95.0% (GC,T) |
Chemical Identifiers
| 937-00-8 | |
| 178.17 | |
| SCURCOWZQJIUGR-UHFFFAOYSA-N | |
| 136751 | |
| FC(F)(F)C1=CC=CC(S)=C1 |
| C7H5F3S | |
| MFCD00041142 | |
| 3-trifluoromethyl thiophenol, 3-trifluoromethyl benzenethiol, 3-trifluoromethyl benzene-1-thiol, 3-trifluoromethylthiophenol, m-trifluoromethylthiophenol, 3-mercaptobenzotrifluoride, m-trifluoromethyl benzenethiol, benzenethiol, 3-trifluoromethyl, pubchem4600 | |
| 3-(trifluoromethyl)benzene-1-thiol |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator