missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzenethiol 95.0+%, TCI America™

Catalog No. p-7137413
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27805G 5 g
T278025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27805G Supplier TCI America Supplier No. T27805G
Only null left

Chemical Identifiers

CAS 937-00-8
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.17
MDL Number MFCD00041142
InChI Key SCURCOWZQJIUGR-UHFFFAOYSA-N
Synonym 3-trifluoromethyl thiophenol, 3-trifluoromethyl benzenethiol, 3-trifluoromethyl benzene-1-thiol, 3-trifluoromethylthiophenol, m-trifluoromethylthiophenol, 3-mercaptobenzotrifluoride, m-trifluoromethyl benzenethiol, benzenethiol, 3-trifluoromethyl, pubchem4600
PubChem CID 136751
IUPAC Name 3-(trifluoromethyl)benzene-1-thiol
SMILES FC(F)(F)C1=CC=CC(S)=C1

Specifications

Color Yellow
Boiling Point 163°C
Quantity 5 g
UN Number 2810
Formula Weight 178.17
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzenethiol
TSCA No
Recommended Storage Refrigerator