missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzaldehyde, 97%

Catalog No. AAA1046322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1046322 100 g
AAA1046306 5 g
AAA1046314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1046322 Supplier Thermo Scientific Chemicals Supplier No. A1046322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 454-89-7 | C8H5F3O | 174.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 454-89-7
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.122
MDL Number MFCD00003373
InChI Key NMTUHPSKJJYGML-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzaldehyde, m-trifluoromethylbenzaldehyde, benzaldehyde, 3-trifluoromethyl, alpha,alpha,alpha-trifluoro-m-tolualdehyde, alpha,alpha,alpha-trifluoro-3-tolualdehyde, unii-at7h4h5u3e, a,a,a-trifluoro-m-tolualdehyde, 3-trifluoromethyl-benzaldehyde, mtf-bad, 3-formylbenzotrifluoride
PubChem CID 67990
IUPAC Name 3-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=CC(=C1)C(F)(F)F)C=O

Specifications

Density 1.304
Boiling Point 83°C to 86°C (30 mmHg)
Flash Point 68°C (154°F)
Odor Almond-like
Refractive Index 1.465
Quantity 100 g
Beilstein 2327537
Sensitivity Air Sensitive
Formula Weight 174.12
Percent Purity 97%
Chemical Name or Material 3- (Trifluoromethyl)benzaldehyde
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 207-228-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only