missing translation for 'onlineSavingsMsg'
Learn More

3-Thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T19751G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19751G Supplier TCI America Supplier No. T19751G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Thiopheneboronic Acid (contains varying amounts of Anhydride)
CAS 6165-69-1
Color White-Yellow
Molecular Formula C4H5BO2S
Quantity 1 g
MDL Number MFCD00151851
Synonym 3-thiopheneboronic acid, thiophene-3-boronic acid, 3-thienylboronic acid, thiophene 3-boronic acid, thiophene-3-boronicacid, thiophen-3-yl-3-boronic acid, 3-thienyl boronic acid, 3-thiophene boronic acid, thiophen-3-yl-boranediol, boronic acid, 3-thienyl
InChI Key QNMBSXGYAQZCTN-UHFFFAOYSA-N
SMILES OB(O)C1=CSC=C1
IUPAC Name (thiophen-3-yl)boronic acid
Molecular Weight (g/mol) 127.95
PubChem CID 581760
Formula Weight 127.95
Physical Form Crystalline Powder
Show More Show Less