missing translation for 'onlineSavingsMsg'
Learn More

3-Tétradécylthiophène 98,0+%, TCI America™

Numéro de catalogue. T31045G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T31045G 5 g
T310425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T31045G Fournisseur TCI America Code fournisseur T31045G

Identifiants chimiques

CAS 110851-66-6
Molecular Formula C18H32S
Molecular Weight (g/mol) 280.51
MDL Number MFCD09750675
InChI Key CAEIOINMYGTXNS-UHFFFAOYSA-N
PubChem CID 14900816
IUPAC Name 3-tétradécylthiophène
SMILES CCCCCCCCCCCCCCC1=CSC=C1

Spécifications

Color Jaune
Quantity 5 g
Formula Weight 280.51
Percent Purity ≥98.0% (GC)
Physical Form Liquide
Chemical Name or Material 3-Tetradecylthiophene