missing translation for 'onlineSavingsMsg'
Learn More

3-Taraxerol, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012891
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012891 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012891 Supplier Merck Emd Millipore Supplier No. 7593410MG
Food and Beverages

Specifications

CAS 127-22-0
Physical Form Neat
Linear Formula C30H50 O
MDL Number MFCD13186859
Synonym (3 beta)-D-Friedoolean-14-en-3-ol; (3 beta13 alpha)-13-Methyl-27-norolean-14-en-3-ol; Alnulin; Skimmiol; Taraxerol; Tiliadin
Type Analytical Standard
Quantity 10 mg
Formula Weight 426.72
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard