missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrrolidinol, 97%

Catalog No. AC425750050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425750050 5 g Glass bottle
AC425750010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425750050 Supplier Thermo Scientific Chemicals Supplier No. 425750050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40499-83-0 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40499-83-0
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005256
InChI Key JHHZLHWJQPUNKB-UHFFFAOYNA-N
Synonym 3-pyrrolidinol, 3-hydroxypyrrolidine, dl-3-pyrrolidinol, 3-hydroxy pyrrolidine, dl-3-hydroxypyrrolidine, rs-3-hydroxypyrrolidine, 3-hydroxypyrrolidinehydrochloride, pyrrolidin-3ol, pyrrolidin-3-o, pyrrolidine-3-ol
PubChem CID 98210
IUPAC Name pyrrolidin-3-ol
SMILES OC1CCNC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0760g/mL
Boiling Point 179°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4881
Quantity 5 g
Specific Gravity 1.076
Formula Weight 87.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Pyrrolidinol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 254-944-5
Recommended Storage Product discolors to brown on storage

RUO – Research Use Only