missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride, 90%, Thermo Scientific™
Chemical Identifiers
| CAS | 887922-93-2 |
|---|---|
| Molecular Formula | C12H16ClNO2 |
| Molecular Weight (g/mol) | 241.715 |
| InChI Key | BORITTZEFSYNFT-UHFFFAOYSA-N |
| Synonym | 3-pyrrolidin-1-ylmethyl benzoic acid hydrochloride, 3-1-pyrrolidylmethyl benzoic acid hydrochloride, 3-pyrrolidinylmethyl benzoic acid, chloride, 3-1-pyrrolidylmethyl benzoicacidhydrochloride, 3-pyrrolidin-1-ylmethyl-benzoic acid hydochloride, 3-pyrrolidin-1-ylmethyl-benzoic acid hydrochloride, 3-pyrrolidin-1-yl methyl benzoic acid-hydrogen chloride 1/1 |
| PubChem CID | 18525874 |
| IUPAC Name | 3-(pyrrolidin-1-ylmethyl)benzoic acid;hydrochloride |
| SMILES | C1CCN(C1)CC2=CC=CC(=C2)C(=O)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
CC55751CB
|
N/A | 250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC55751DA
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC55751DE
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 887922-93-2 | |
| 241.715 | |
| 3-pyrrolidin-1-ylmethyl benzoic acid hydrochloride, 3-1-pyrrolidylmethyl benzoic acid hydrochloride, 3-pyrrolidinylmethyl benzoic acid, chloride, 3-1-pyrrolidylmethyl benzoicacidhydrochloride, 3-pyrrolidin-1-ylmethyl-benzoic acid hydochloride, 3-pyrrolidin-1-ylmethyl-benzoic acid hydrochloride, 3-pyrrolidin-1-yl methyl benzoic acid-hydrogen chloride 1/1 | |
| 3-(pyrrolidin-1-ylmethyl)benzoic acid;hydrochloride |
| C12H16ClNO2 | |
| BORITTZEFSYNFT-UHFFFAOYSA-N | |
| 18525874 | |
| C1CCN(C1)CC2=CC=CC(=C2)C(=O)O.Cl |
Specifications
| 887922-93-2 | |
| Amber Glass Bottle | |
| 3-pyrrolidin-1-ylmethyl benzoic acid hydrochloride, 3-1-pyrrolidylmethyl benzoic acid hydrochloride, 3-pyrrolidinylmethyl benzoic acid, chloride, 3-1-pyrrolidylmethyl benzoicacidhydrochloride, 3-pyrrolidin-1-ylmethyl-benzoic acid hydochloride, 3-pyrrolidin-1-ylmethyl-benzoic acid hydrochloride, 3-pyrrolidin-1-yl methyl benzoic acid-hydrogen chloride 1/1 | |
| C1CCN(C1)CC2=CC=CC(=C2)C(=O)O.Cl | |
| 241.715 | |
| 241.72 | |
| 3-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride |
| 90% | |
| C12H16ClNO2 | |
| BORITTZEFSYNFT-UHFFFAOYSA-N | |
| 3-(pyrrolidin-1-ylmethyl)benzoic acid;hydrochloride | |
| 18525874 | |
| 90% |
Safety and Handling
Warning