missing translation for 'onlineSavingsMsg'
Learn More

3-(Pyrrolidin-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 884507-42-0
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.26
MDL Number MFCD07782420
InChI Key XHHOLQHPKIDQEM-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-ylmethyl benzaldehyde, 3-pyrrolidin-1-yl methyl benzaldehyde, benzaldehyde, 3-1-pyrrolidinylmethyl, 1-3-formylbenzyl pyrrolidine, 3-pyrrolidinylmethyl benzaldehyde, 3-pyrrolidin-1-ylmethyl-benzaldehyde
PubChem CID 18525870
IUPAC Name 3-(pyrrolidin-1-ylmethyl)benzaldehyde
SMILES O=CC1=CC(CN2CCCC2)=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC55704CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC55704DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

884507-42-0
189.26
XHHOLQHPKIDQEM-UHFFFAOYSA-N
18525870
O=CC1=CC(CN2CCCC2)=CC=C1
C12H15NO
MFCD07782420
3-pyrrolidin-1-ylmethyl benzaldehyde, 3-pyrrolidin-1-yl methyl benzaldehyde, benzaldehyde, 3-1-pyrrolidinylmethyl, 1-3-formylbenzyl pyrrolidine, 3-pyrrolidinylmethyl benzaldehyde, 3-pyrrolidin-1-ylmethyl-benzaldehyde
3-(pyrrolidin-1-ylmethyl)benzaldehyde

Specifications

884507-42-0
100.0
C12H15NO
3-pyrrolidin-1-ylmethyl benzaldehyde, 3-pyrrolidin-1-yl methyl benzaldehyde, benzaldehyde, 3-1-pyrrolidinylmethyl, 1-3-formylbenzyl pyrrolidine, 3-pyrrolidinylmethyl benzaldehyde, 3-pyrrolidin-1-ylmethyl-benzaldehyde
O=CC1=CC(CN2CCCC2)=CC=C1
189.26
189.26
3-(Pyrrolidin-1-ylmethyl)benzaldehyde
96.5
97%
MFCD07782420
XHHOLQHPKIDQEM-UHFFFAOYSA-N
3-(pyrrolidin-1-ylmethyl)benzaldehyde
18525870
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents