missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrrolidin-1-ylaniline, 97%, Thermo Scientific™

Catalog No. CC38514DA
Change view
Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC38514DA 1g Amber Glass Bottle
CC38514EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC38514DA Supplier Thermo Scientific Chemicals Supplier No. CC38514DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 115833-93-7
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
InChI Key DNPBFTQZONVQDX-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-yl aniline, 3-pyrrolidin-1-yl-phenylamine, 3-1-pyrrolidinyl aniline, benzenamine,3-1-pyrrolidinyl, benzenamine, 3-1-pyrrolidinyl, 3-pyrrolidinylphenylamine, 3-pyrrolizinoaniline, acmc-20e1pl, 1-3-aminophenyl pyrrolidine
PubChem CID 1518808
IUPAC Name 3-pyrrolidin-1-ylaniline
SMILES C1CCN(C1)C2=CC(=CC=C2)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 3-Pyrrolidin-1-ylaniline