missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-5-ylbenzoic acid, ≥97%, Thermo Scientific™

Numéro de catalogue. CC41701DE
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC41701DE 5g
CC41701CB 250mg
CC41701DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC41701DE Fournisseur Thermo Scientific Chemicals Code fournisseur CC41701DE

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 852180-74-6
Formule moléculaire C11H8N2O2
Poids moléculaire (g/mol) 200.197
Numéro MDL MFCD05864810
Clé InChI URQRFVRGUHUTMI-UHFFFAOYSA-N
Synonyme 3-pyrimidin-5-yl-benzoic acid, 3-pyrimidin-5-yl benzoic acid, 3-5-pyrimidinyl benzoic acid, benzoic acid,3-5-pyrimidinyl, benzoic acid, 3-5-pyrimidinyl, 3-pyrimidin-5-yl-benzoicacid, benzoic acid, 3-5-pyrimidinyl-9ci
PubChem CID 4187250
Nom de l’IUPAC 3-pyrimidin-5-ylbenzoic acid
SOURIRES C1=CC(=CC(=C1)C(=O)O)C2=CN=CN=C2

Spécifications

Nom ou substance chimique 3-Pyrimidin-5-ylbenzoic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Quantité 5g
Poids de formule 200.2
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.