missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-5-ylbenzaldehyde, 97%, Thermo Scientific™

Numéro de catalogue. p-3485523
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC41704CB 250mg
CC41704DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC41704CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC41704CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 640769-70-6
Formule moléculaire C11H8N2O
Poids moléculaire (g/mol) 184.198
Numéro MDL MFCD05864833
Clé InChI RAHJVJBPRWJPHE-UHFFFAOYSA-N
Synonyme 3-pyrimidin-5-yl benzaldehyde, 3-pyrimidin-5-yl-benzaldehyde, 3-5-pyrimidinyl benzaldehyde, benzaldehyde, 3-5-pyrimidinyl, acmc-1b2vh, benzaldehyde,3-5-pyrimidinyl
PubChem CID 4187249
Nom de l’IUPAC 3-pyrimidin-5-ylbenzaldehyde
SOURIRES C1=CC(=CC(=C1)C=O)C2=CN=CN=C2

Spécifications

Nom ou substance chimique 3-Pyrimidin-5-ylbenzaldehyde
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Quantité 250mg
Poids de formule 184.2
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.