missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. P18605G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18605G Supplier TCI America Supplier No. P18605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent used as a building block in organic synthesis.

Chemical Identifiers

CAS 107658-27-5
Molecular Formula C6H4F3NO3S
Molecular Weight (g/mol) 227.157
MDL Number MFCD00799579
InChI Key ZRNDSLLIAOLRJS-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 3-Pyridyl Ester, 3-Pyridyl Triflate
PubChem CID 11031579
IUPAC Name pyridin-3-yl trifluoromethanesulfonate
SMILES C1=CC(=CN=C1)OS(=O)(=O)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 227.16
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 3-Pyridyl Trifluoromethanesulfonate
TSCA No
Recommended Storage Refrigerator