missing translation for 'onlineSavingsMsg'
Learn More

3-(Pyrid-2-yloxy)benzoic acid, 97%, Thermo Scientific™

Catalog No. CC50201CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC50201CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC50201CB Supplier Thermo Scientific Chemicals Supplier No. CC50201CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51362-30-2
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD09025853
InChI Key LYSIEAIIZBZRCE-UHFFFAOYSA-N
Synonym 3-pyridin-2-yloxy benzoic acid, 3-pyrid-2-yloxy benzoic acid, 3-2-pyridyloxy benzoic acid, 3-2 pyridyloxy-benzoic acid, 3-pyridin-2-yl oxy benzoic acid, benzoic acid, 3-2-pyridinyloxy
PubChem CID 21901423
IUPAC Name 3-pyridin-2-yloxybenzoic acid
SMILES C1=CC=NC(=C1)OC2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 215.21
Percent Purity 95%
Chemical Name or Material 3-(Pyrid-2-yloxy)benzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.