Learn More
BeLLCo Glass Material: 316l SST with electropolish finish, ra 32 or better, inside & out Dimensions: harvest tube (3/8"" od) - 1"" above slug, 15"" insertion length Addition tube (1/4""od) - 1"" above slug, 2 21/64"" below Vent tube (1/4"" od) - 1"" above slug
Numéro de catalogue. 501944851
Encompass
Material: 316l SST with electropolish finish, ra 32 or better, inside & out Dimensions: harvest tube (3/8" od) - 1" above slug, 15" insertion length Addition tube (1/4"od) - 1" above slug, 2 21/64" below Vent tube (1/4" od) - 1" above slug
Numéro de catalogue. 501944851 Fournisseur BeLLCo Glass Code fournisseur 196463008
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Material: 316l SST with electropolish finish, ra 32 or better, inside & out Dimensions: harvest tube (3/8" od) - 1" above slug, 15" insertion length Addition tube (1/4"od) - 1" above slug, 2 21/64" below Vent tube (1/4" od) - 1" above slug