missing translation for 'onlineSavingsMsg'
Learn More

(3-Pipéridinophényl)méthanol, 97%, Thermo Scientific™

Numéro de catalogue. CC34009DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC34009DA 1 g Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC34009DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC34009DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 852180-58-6
Formule moléculaire C12H17NO
Poids moléculaire (g/mol) 191.274
Numéro MDL MFCD07772816
Clé InChI HNESBVGXCDQHLJ-UHFFFAOYSA-N
Synonyme 3-piperidinophenyl methanol, 3-piperidin-1-yl phenyl methanol, 3-piperidin-1-ylphenyl methanol, 3-piperidylphenyl methan-1-ol, benzenemethanol,3-1-piperidinyl
PubChem CID 7162330
Nom de l’IUPAC (3-pipéridin-1-ylphényl)méthanol
SOURIRES C1CCN(CC1)C2=CC=CC(=C2)CO

Spécifications

Nom ou substance chimique (3-Piperidinophenyl)methanol
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 191.27
Pourcentage de pureté 97%
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.