missing translation for 'onlineSavingsMsg'
Learn More

3-Phosphoshikimic Acid lithium salt, 95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015032
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015032 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015032 Supplier Merck Emd Millipore Supplier No. 5538510MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63959-45-5
Molecular Formula C7H11O8P
Molecular Weight (g/mol) 254.13
MDL Number MFCD04039676
InChI Key QYOJSKGCWNAKGW-PBXRRBTRSA-N
Synonym Lithium 3-phosphoshikimate; Lithium shikimate-3-phosphate; S-3-P; S3P; Shikimic acid-3-phosphate lithium salt
IUPAC Name (3R,4S,5R)-4,5-dihydroxy-3-(phosphonooxy)cyclohex-1-ene-1-carboxylic acid
SMILES O[C@@H]1CC(=C[C@@H](OP(O)(O)=O)[C@H]1O)C(O)=O

Specifications

Linear Formula C7H11O8P · xLi+
Quantity 10 mg
Formula Weight 254.13 (free acid basis)
Percent Purity ≥95.0% (HPLC)