missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenylthio)thiophene, 97%

Catalog No. AAA1258506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1258506 5 g
AAA1258503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1258506 Supplier Thermo Scientific Chemicals Supplier No. A1258506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16718-11-9 | C10H8S2 | 192.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16718-11-9
Molecular Formula C10H8S2
Molecular Weight (g/mol) 192.29
MDL Number MFCD00052043
InChI Key WQYNBKIZHCGYCV-UHFFFAOYSA-N
Synonym 3-phenylthio thiophene, 3-phenylsulfanyl thiophene, thiophene,3-phenylthio, 3-phenylthiothiophene, acmc-1bt9i, 3-phenylsulphanyl thiophene, phenyl-3-thienyl-sulphide
PubChem CID 2778902
IUPAC Name 3-phenylsulfanylthiophene
SMILES S(C1=CSC=C1)C1=CC=CC=C1

Specifications

Boiling Point 111°C to 112°C (0.6 mmHg)
Quantity 5 g
Formula Weight 192.3
Percent Purity 97%
Chemical Name or Material 3-(Phenylthio)thiophene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only