missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenylsulfonyl)propionitrile, 99%

Catalog No. AAL2019614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2019614 25 g
AAL2019606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2019614 Supplier Thermo Scientific Chemicals Supplier No. L2019614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10154-75-3 | C9H9NO2S | 195.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10154-75-3
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.236
MDL Number MFCD00047798
InChI Key OVZPZPVSUAKTCM-UHFFFAOYSA-N
Synonym 3-phenylsulfonyl propionitrile, 3-phenylsulfonyl propanenitrile, 3-phenylsulphonyl propiononitrile, 3-benzenesulfonyl propanenitrile, 3-phenylsulfonylpropionitrile, acmc-1bvrh, enamine_005271, 3-phenylsulfonyl propanitrile, 3-phenylsulphonyl-propionitrile, propanenitrile, 3-phenylsulfonyl
PubChem CID 82413
IUPAC Name 3-(benzenesulfonyl)propanenitrile
SMILES C1=CC=C(C=C1)S(=O)(=O)CCC#N

Specifications

Melting Point 94°C to 96°C
Odor Odorless
Quantity 25 g
Beilstein 2051019
Formula Weight 195.24
Percent Purity 99%
Chemical Name or Material 3-(Phenylsulfonyl)propionitrile
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 233-424-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only