missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpyridine 94.0+%, TCI America™

Catalog No. P10405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10405G 5 g
P104025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10405G Supplier TCI America Supplier No. P10405G

Chemical Identifiers

CAS 1008-88-4
Molecular Formula C11H9N
Molecular Weight (g/mol) 155.2
MDL Number MFCD00006380
InChI Key HJKGBRPNSJADMB-UHFFFAOYSA-N
Synonym pyridine, 3-phenyl, 3-phenyl-pyridine, m-phenylpyridine, unii-edz6kb7jvc, 5-phenylpyridine, 3-phenyipyridine, edz6kb7jvc, 3-azabiphenyl, 3-pyridylbenzene, 3-phenyl pyridine
PubChem CID 13886
IUPAC Name 3-phenylpyridine
SMILES C1=CC=C(C=C1)C2=CN=CC=C2

Specifications

Boiling Point 274°C
Quantity 5 g
Formula Weight 155.20
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenylpyridine