missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpyridine, 95%

Catalog No. AAB2393006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2393006 5 g
AAB2393014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2393006 Supplier Thermo Scientific Chemicals Supplier No. B2393006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1008-88-4 | C11H9N | 155.2 g/mol

3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.

Chemical Identifiers

CAS 1008-88-4
Molecular Formula C11H9N
Molecular Weight (g/mol) 155.2
MDL Number MFCD00006380
InChI Key HJKGBRPNSJADMB-UHFFFAOYSA-N
Synonym pyridine, 3-phenyl, 3-phenyl-pyridine, m-phenylpyridine, unii-edz6kb7jvc, 5-phenylpyridine, 3-phenyipyridine, edz6kb7jvc, 3-azabiphenyl, 3-pyridylbenzene, 3-phenyl pyridine
PubChem CID 13886
IUPAC Name 3-phenylpyridine
SMILES C1=CC=C(C=C1)C2=CN=CC=C2

Specifications

Density 1.085
Boiling Point 272°C
Flash Point >110°C (230°F)
Refractive Index 1.616
Quantity 5 g
Beilstein 110400
Solubility Information Soluble in chloroform,dichloromethane and ethyl acetate.
Formula Weight 155.2
Percent Purity 97%
Chemical Name or Material 3-Phenylpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-762-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only