missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropylamine, 98+%

Catalog No. AAL0302309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0302309 10 g
AAL0302318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0302309 Supplier Thermo Scientific Chemicals Supplier No. L0302309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2038-57-5 | C9H13N | 135.21 g/mol

3-Phenylpropylamine is is a compound used for the synthesis of various pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpropylamine is is a compound used for the synthesis of various pharmaceuticals.

Solubility
Soluble in Chloroform, Ethyl Acetate. Not miscible in water.

Notes
Store away from air and under dry inert gas. Air sensitive. Incompatible with acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide.

Chemical Identifiers

CAS 2038-57-5
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008224
InChI Key LYUQWQRTDLVQGA-UHFFFAOYSA-N
Synonym 3-phenylpropylamine, benzenepropanamine, 1-amino-3-phenylpropane, hydrocinnamylamine, gamma-phenylpropylamine, 3-phenyl-n-propylamine, 3-phenyl-1-propylamine, 3-phenyl-1-propanamine, propylamine, 3-phenyl, 3-phenylpropanamine
PubChem CID 16259
IUPAC Name 3-phenylpropan-1-amine
SMILES NCCCC1=CC=CC=C1

Specifications

Density 0.949
Boiling Point 220°C to 221°C
Flash Point 90°C (194°F)
Odor Amine-like
Refractive Index 1.525
Quantity 10 g
UN Number UN2735
Beilstein 2205526
Sensitivity Air Sensitive
Solubility Information Soluble in Chloroform,Ethyl Acetate. Not miscible in water.
Formula Weight 135.21
Percent Purity ≥98%
Chemical Name or Material 3-Phenylpropylamine
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 218-012-1
RTECSNumber UI4590000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only