missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionic acid, 99%

Numéro de catalogue. p-7022232
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1490822 100 g
AAA1490836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAA1490822 Fournisseur Thermo Scientific Chemicals Code fournisseur A1490822
Il en reste null

CAS: 501-52-0 | C9H10O2 | 150.177 g/mol

Hydrocinnamic acid is inhibitor of carboxypeptidase. Hydrocinnamic acid was used in the synthesis of three medium-chain acyl-Coenzyme A?s i.e. 3-phenylpropionyl-CoA from mixed anhydrides of fatty acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrocinnamic acid is inhibitor of carboxypeptidase. Hydrocinnamic acid was used in the synthesis of three medium-chain acyl-Coenzyme A′s i.e. 3-phenylpropionyl-CoA from mixed anhydrides of fatty acids.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Identifiants chimiques

CAS 501-52-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002771
InChI Key XMIIGOLPHOKFCH-UHFFFAOYSA-N
Synonym hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid
PubChem CID 107
ChEBI CHEBI:28631
IUPAC Name 3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CCC(=O)O

Spécifications

Melting Point 47°C to 50°C
Density 1.071
Boiling Point 280°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 907515
Merck Index 14,4784
Solubility Information Soluble in water
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material 3-Phenylpropionic acid
Afficher plus de résultats Afficher moins de résultats
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 207-924-5
RTECSNumber DA8600000
TSCA Yes

RUO – Research Use Only