missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propyne, 97%, stab.

Numéro de catalogue. p-7035773
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAH5350006 5 g
AAH5350014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAH5350006 Fournisseur Thermo Scientific Chemicals Code fournisseur H5350006
Il en reste null

CAS: 10147-11-2 | C9H8 | 116.163 g/mol

3-Phenyl-1-propyne is used as a starting material in the synthesis of 4-phenyl-2-butyn-1-ol. It is also used in the preparation of beta-hydroxytriazoles and indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenyl-1-propyne is used as a starting material in the synthesis of 4-phenyl-2-butyn-1-ol. It is also used in the preparation of beta-hydroxytriazoles and indoles.

Notes
Incompatible with acids and bases.

Identifiants chimiques

CAS 10147-11-2
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
MDL Number MFCD00134431
InChI Key NGKSKVYWPINGLI-UHFFFAOYSA-N
Synonym 3-phenyl-1-propyne, 2-propynyl benzene, benzylacetylene, prop-2-yn-1-ylbenzene, benzene, 2-propynyl, propargylbenzol, propargylbenzene, benzyl acetylene, 3-phenyl-propyne, 3-phenyl-1-propyn
PubChem CID 575753
IUPAC Name prop-2-ynylbenzene
SMILES C#CCC1=CC=CC=C1

Spécifications

Density 0.934
Boiling Point 75°C (20 mmHg)
Flash Point 52°C (126°F)
Refractive Index 1.526
Linear Formula C6H5CH2C≡CH
Quantity 5 g
UN Number UN3295
Formula Weight 116.16
Percent Purity 97%
Chemical Name or Material 3-Phenyl-1-propyne, stabilized
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only