missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxypropionic Acid 98.0+%, TCI America™

Catalog No. P14005G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14005G Supplier TCI America Supplier No. P14005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7170-38-9
Molecular Formula C9H9O3
Molecular Weight (g/mol) 165.17
MDL Number MFCD00002769
InChI Key BUSOTUQRURCMCM-UHFFFAOYSA-M
Synonym 3-phenoxypropionic acid, propanoic acid, 3-phenoxy, beta-phenoxypropionic acid, 3-penoxypropionic acid, 3-phenoxy-propionic acid, 3-phenoxypropionicacid, b-phenoxypropionate, 3-phenoxypropionate, 3-phenoxy-propanoate, beta-phenoxypropionate
PubChem CID 81596
IUPAC Name 3-phenoxypropanoate
SMILES [O-]C(=O)CCOC1=CC=CC=C1

Specifications

Melting Point 98°C
Color White
Boiling Point 245°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenoxypropionic Acid