missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Phenoxymethyl)benzoic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 31719-75-2 |
|---|---|
| Molecular Formula | C14H12O3 |
| Molecular Weight (g/mol) | 228.247 |
| InChI Key | URLYREZIPSJJQU-UHFFFAOYSA-N |
| Synonym | 3-phenoxymethyl benzoic acid, benzoic acid,3-phenoxymethyl, 4rg |
| PubChem CID | 3729884 |
| IUPAC Name | 3-(phenoxymethyl)benzoic acid |
| SMILES | C1=CC=C(C=C1)OCC2=CC=CC(=C2)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
CC63601CB
|
Thermo Scientific Chemicals
CC63601CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC63601DA
|
Thermo Scientific Chemicals
CC63601DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC63601DE
|
Thermo Scientific Chemicals
CC63601DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 31719-75-2 | |
| 228.247 | |
| 3-phenoxymethyl benzoic acid, benzoic acid,3-phenoxymethyl, 4rg | |
| 3-(phenoxymethyl)benzoic acid |
| C14H12O3 | |
| URLYREZIPSJJQU-UHFFFAOYSA-N | |
| 3729884 | |
| C1=CC=C(C=C1)OCC2=CC=CC(=C2)C(=O)O |
Specifications
| 31719-75-2 | |
| Amber Glass Bottle | |
| 250mg | |
| URLYREZIPSJJQU-UHFFFAOYSA-N | |
| 3-(phenoxymethyl)benzoic acid | |
| 3729884 | |
| 97% |
| 97% | |
| C14H12O3 | |
| 3-phenoxymethyl benzoic acid, benzoic acid,3-phenoxymethyl, 4rg | |
| C1=CC=C(C=C1)OCC2=CC=CC(=C2)C(=O)O | |
| 228.247 | |
| 228.25 | |
| 3-(phenoxymethyl)benzoic acid |
Safety and Handling
GHS Signal Word: Warning