missing translation for 'onlineSavingsMsg'
Learn More

3-Penten-2-one, tech. 85%

Catalog No. AAL1303106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1303106 5 g
AAL1303103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1303106 Supplier Thermo Scientific Chemicals Supplier No. L1303106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 625-33-2 | C5H8O | 84.118 g/mol

3-Penten-2-one

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Penten-2-one

Solubility
Slightly soluble in Water

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with bases, reducing agents and oxidizing agents.

Chemical Identifiers

CAS 625-33-2
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00009290
InChI Key LABTWGUMFABVFG-ONEGZZNKSA-N
Synonym z-pent-3-en-2-one, unii-w8xx21whyc, z-3-penten-2-one, w8xx21whyc, 3-penten-2-one, 3z-pent-3-en-2-one, 3-penten-2-one, z, cis-3-penten-2-one, 3-penten-2-one, cis
PubChem CID 637920
IUPAC Name (E)-pent-3-en-2-one
SMILES CC=CC(=O)C

Specifications

Density 0.862
Boiling Point 119°C to 121°C
Flash Point 21°C (69°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.4355
Quantity 5 g
UN Number UN1992
Beilstein 1633505
Solubility Information Slightly soluble in Water
Formula Weight 84.12
Percent Purity 85%
Grade Technical
Chemical Name or Material 3-Penten-2-one
Show More Show Less
Hazard Category H225-H312-H315-H319-H335
Hazard Statement GHS H Statement
H225-H311-H315-H319-H335
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 210-888-3
RTECSNumber SB3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only