missing translation for 'onlineSavingsMsg'
Learn More

3-Pentanone 98.0+%, TCI America™

Catalog No. P006125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P006125ML 25 mL
P0061500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P006125ML Supplier TCI America Supplier No. P006125ML

Chemical Identifiers

CAS 96-22-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00009320
InChI Key FDPIMTJIUBPUKL-UHFFFAOYSA-N
Synonym 3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone
PubChem CID 7288
ChEBI CHEBI:67886
IUPAC Name pentan-3-one
SMILES CCC(=O)CC

Specifications

Color Colorless
Boiling Point 102°C
Quantity 25 mL
UN Number 1156
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Pentanone
EINECSNumber (2)-2474
RTECSNumber SA8050000
TSCA Yes