missing translation for 'onlineSavingsMsg'
Learn More

3-Oxocyclopentanecarboxylic acid, 97%

Catalog No. AAH6041403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6041403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6041403 Supplier Thermo Scientific Chemicals Supplier No. H6041403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-78-2 | C6H8O3 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-78-2
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD01320173
InChI Key RDSNBKRWKBMPOP-UHFFFAOYNA-N
Synonym 3-oxocyclopentanecarboxylic acid, 3-oxo-cyclopentane-carboxylic acid, 3-oxo cyclopentane 1-carboxylic acid, 3-oxo-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-oxo, 3-carboxycyclopentanone, cyclopentanone-3-carboxylic acid, 3-oxo-cyclopentanecarboxylic acid, 3-ketocyclopentanecarboxylic acid, 3-cyclopentanone-1 r-carboxylic acid
PubChem CID 227798
ChEBI CHEBI:71213
IUPAC Name 3-oxocyclopentane-1-carboxylic acid
SMILES OC(=O)C1CCC(=O)C1

Specifications

Melting Point 59°C to 62°C
Quantity 1 g
Formula Weight 128.13
Percent Purity 97%
Chemical Name or Material 3-Oxocyclopentanecarboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only