missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CC46803CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC46803CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC46803CB Supplier Thermo Scientific Chemicals Supplier No. CC46803CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31794-45-3
Molecular Formula C8H6ClNO4S
Molecular Weight (g/mol) 247.649
MDL Number MFCD05664887
InChI Key CGTCULUUVYBAPX-UHFFFAOYSA-N
Synonym 3-oxo-3,4-dihydro-2h-1,4-benzoxazine-6-sulfonyl chloride, 2h-1,4-benzoxazine-6-sulfonyl chloride, 3,4-dihydro-3-oxo, 3-oxo-3,4-dihydro-2h-benzo b 1,4 oxazine-6-sulfonyl chloride, 3,4-dihydro-3-oxo-2h-1,4-benzoxazine-6-sulphonyl chloride, 3-oxo-3,4-dihydro-2h-benzo 1,4 oxazine-6-sulfonyl chloride, 3-oxo-2,4-dihydro-1,4-benzoxazine-6-sulfonyl chloride, 6-chlorosulfonyl-2h,4h-benzo e 1,4-oxazin-3-one, 6-chlorosulphonyl-3-oxo-1,4-benzoxazine, 3-oxo-3,4-dihydro-2h-1,4-benzooxazine-6-sulfonyl chloride, 3-oxo-3,4-dihydro-2h-1,4-benzoxazine-6-sulfonyl ch
PubChem CID 5200229
IUPAC Name 3-oxo-4H-1,4-benzoxazine-6-sulfonyl chloride
SMILES C1C(=O)NC2=C(O1)C=CC(=C2)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 247.66
Percent Purity 97%
Chemical Name or Material 3-Oxo-3, 4-dihydro-2H-1, 4-benzoxazine-6-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.