missing translation for 'onlineSavingsMsg'
Learn More

3-Nitropyrazole 98.0+%, TCI America™

Catalog No. N09375G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09375G 5 g
N09371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09375G Supplier TCI America Supplier No. N09375G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26621-44-3
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00159621,MFCD00238787
InChI Key MZRUFMBFIKGOAL-UHFFFAOYSA-N
Synonym 3-nitro-1h-pyrazole, 3-nitropyrazole, 1h-pyrazole, 3-nitro, 3-nitro-2h-pyrazole, nitropyrazole, pyrazole, 3-nitro, 5-nitro-1h-pyrazol, djw 94, djw-94, 5-nitropyrazole
PubChem CID 123419
IUPAC Name 5-nitro-1H-pyrazole
SMILES [O-][N+](=O)C1=CC=NN1

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 5 g
Formula Weight 113.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitropyrazole