missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenyl isocyanate, 97%

Catalog No. AAA1692006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1692006 5 g
AAA1692014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1692006 Supplier Thermo Scientific Chemicals Supplier No. A1692006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3320-87-4 | C7H4N2O3 | 164.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3320-87-4
Molecular Formula C7H4N2O3
Molecular Weight (g/mol) 164.12
MDL Number MFCD00007220
InChI Key GFFGYTMCNVMFAJ-UHFFFAOYSA-N
Synonym 3-nitrophenyl isocyanate, m-nitrophenyl isocyanate, benzene, 1-isocyanato-3-nitro, 3-nitrophenylisocyanate, isocyanic acid, m-nitrophenyl ester, 3-nitrobenzenisocyanate, m-nitrophenylisocyanate, 3-nitropheny isocyanate, pubchem15729, acmc-1ckin
PubChem CID 76835
IUPAC Name 1-isocyanato-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(=C1)N=C=O

Specifications

Melting Point 51°C to 53°C
Boiling Point 130°C to 131°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Quantity 5 g
UN Number UN2811
Sensitivity Moisture sensitive
Formula Weight 164.12
Percent Purity 97%
Chemical Name or Material 3-Nitrophenyl isocyanate
Hazard Category H302+H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 222-025-8
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only