missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrochalcone 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 144°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 253.26 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Nitrochalcone |
Sécurité et manipulation
| RTECSNumber | FL7075490 |
| TSCA | No |