missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzoyl chloride, 98%

Catalog No. AAA1057930
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1057930 250 g
AAA1057918 50 g
AAA105790B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1057930 Supplier Thermo Scientific Chemicals Supplier No. A1057930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121-90-4 | C7H4ClNO3 | 185.563 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-90-4
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.563
MDL Number MFCD00007247
InChI Key NXTNASSYJUXJDV-UHFFFAOYSA-N
Synonym m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid
PubChem CID 8495
IUPAC Name 3-nitrobenzoyl chloride
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl

Specifications

Melting Point 31°C to 35°C
Density 1.42
Boiling Point 276°C to 278°C (decomposition)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 250 g
UN Number UN3261
Beilstein 777186
Sensitivity Moisture sensitive
Formula Weight 185.57
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzoyl chloride
Show More Show Less
Hazard Category H311-H314-H335
Hazard Statement GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 204-505-9
RTECSNumber DM6646000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only