missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzhydrazide, 98+%

Catalog No. AAB2080014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2080014 25 g
AAB2080006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2080014 Supplier Thermo Scientific Chemicals Supplier No. B2080014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 618-94-0 | C7H7N3O3 | 181.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-94-0
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00017059
InChI Key NQEWXLVDAVTOHM-UHFFFAOYSA-N
Synonym 3-nitrobenzhydrazide, 3-nitrobenzoylhydrazine, m-nitrobenzhydrazide, 3-nitrobenzoic acid hydrazide, benzoic acid, 3-nitro-, hydrazide, m-nitrobenzoylhydrazide, m-nitrobenzoylhydrazine, m-nitrobenzoic hydrazide, m-nitrobenzoic acid hydrazide, benzoic acid, m-nitro-, hydrazide
PubChem CID 3291498
IUPAC Name 3-nitrobenzohydrazide
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)NN

Specifications

Melting Point 152°C to 155°C
Quantity 25 g
Beilstein 645512
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material 3-Nitrobenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-572-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only