missing translation for 'onlineSavingsMsg'
Learn More

3-Nitroaniline Hydrochloride 98.0+%, TCI America™

Catalog No. N012025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N012025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N012025G Supplier TCI America Supplier No. N012025G
Only null left

Chemical Identifiers

CAS 33240-96-9
Molecular Formula C6H7ClN2O2
Molecular Weight (g/mol) 174.584
MDL Number MFCD00050693
InChI Key FWUUMDDHHPVSPL-UHFFFAOYSA-N
PubChem CID 12215133
IUPAC Name 3-nitroaniline;hydrochloride
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])N.Cl

Specifications

Color Yellow
Quantity 25 g
Formula Weight 174.58
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitroaniline Hydrochloride
EINECSNumber (3)-0392
RTECSNumber BY7370000
TSCA No