missing translation for 'onlineSavingsMsg'
Learn More

3-[N,N-Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulfonic Acid 98.0+%, TCI America™

Catalog No. B149425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B149425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B149425G Supplier TCI America Supplier No. B149425G

Chemical Identifiers

CAS 68399-80-4
Molecular Formula C7H17NO6S
Molecular Weight (g/mol) 243.274
MDL Number MFCD00038353
InChI Key XCBLFURAFHFFJF-UHFFFAOYSA-N
Synonym dipso, 3-bis 2-hydroxyethyl amino-2-hydroxypropane-1-sulfonic acid, 3-n,n-bis 2-hydroxyethyl amino-2-hydroxypropanesulfonic acid, 3-n,n-bis 2-hydroxyethyl amino-2-hydroxy-1-propanesulfonic acid, 1-propanesulfonic acid, 3-bis 2-hydroxyethyl amino-2-hydroxy, 3-bis 2-hydroxyethyl amino-2-hydroxypropanesulfonic acid, 3-bis 2-hydroxyethyl amino-2-hydroxy-1-propanesulfonic acid, n,n-bis 2-hydroxyethyl-3-amino-2-hydroxypropanesulfonic acid
PubChem CID 100210
IUPAC Name 3-[bis(2-hydroxyethyl)amino]-2-hydroxypropane-1-sulfonic acid
SMILES C(CO)N(CCO)CC(CS(=O)(=O)O)O

Specifications

Melting Point 210°C
Color White
Quantity 25 g
Formula Weight 243.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-[N,N-Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulfonic Acid