missing translation for 'onlineSavingsMsg'
Learn More

(3-Morpholinophenyl)methanol, 97%, Thermo Scientific™

Numéro de catalogue. CC33809CB
Modifier l'affichage
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
CC33809CB Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC33809CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC33809CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Spécifications

Nom ou substance chimique (3-Morpholinophenyl)methanol
CAS 145127-38-4
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Formule moléculaire C11H15NO2
Quantité 250mg
Numéro MDL MFCD07772812
Synonyme 3-morpholinophenyl methanol, 3-morpholinobenzyl alcohol, 3-morpholin-4-yl phenyl methanol, 3-morpholin-4-ylphenyl methanol, benzenemethanol,3-4-morpholinyl, benzenemethanol, 3-4-morpholinyl, 3-morpholinobenzylalcohol, acmc-20n4f1, 3-morpholin-4-ylphenyl methan-1-ol
Clé InChI MXBVALXTJZMIJB-UHFFFAOYSA-N
SOURIRES OCC1=CC(=CC=C1)N1CCOCC1
Nom de l’IUPAC (3-morpholin-4-ylphenyl)methanol
Poids moléculaire (g/mol) 193.25
PubChem CID 7162074
Poids de formule 193.24
Pourcentage de pureté 97%
Afficher plus de résultats Afficher moins de résultats
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.