missing translation for 'onlineSavingsMsg'
Learn More

3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate, 97%, Thermo Scientific™

Numéro de catalogue. CC40151DA
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC40151DA 1g
CC40151CB 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC40151DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC40151DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 137605-80-2
Formule moléculaire C12H16ClNO3
Poids moléculaire (g/mol) 257.714
Numéro MDL MFCD09064949
Clé InChI UOMMCPQFAXOMMO-UHFFFAOYSA-N
Synonyme 3-morpholin-4-ylmethyl benzoic acid hydrochloride, 3-morpholinomethyl benzoic acid hydrochloride, 3-morpholin-4-ylmethyl benzoic acid, chloride, 3-morpholin-4-ylmethyl benzoic acid,hydrochloride, 3-morpholin-4-yl methyl benzoic acid hydrochloride, 3-morpholin-4-yl methyl benzoic acid-hydrogen chloride 1/1
PubChem CID 24229486
Nom de l’IUPAC 3-(morpholin-4-ylmethyl)benzoic acid;hydrochloride
SOURIRES C1COCCN1CC2=CC=CC(=C2)C(=O)O.Cl

Spécifications

Nom ou substance chimique 3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate
CAS % minimum 96.5
CAS % maximal 100.0
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 275.74
Pourcentage de pureté 97%
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.