missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene-2-carboxaldehyde 85.0+%, TCI America™

Catalog No. M089925G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M089925G 25 g
M0899100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M089925G Supplier TCI America Supplier No. M089925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5834-16-2
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.173
MDL Number MFCD00005430
InChI Key BSQKBHXYEKVKMN-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarboxaldehyde, 3-methylthiophene-2-carboxaldehyde, 2-formyl-3-methylthiophene, 3-methylthiophene-2-aldehyde, 3-methyl-2-thiophenecarbaldehyde, 3-methyl-2-formylthiophene, 3-methyl-2-thiophene carboxaldehyde, 2-thiophenecarboxaldehyde, 3-methyl, thiophenecarboxaldehyde, methyl, 3-methyl-thiophene-2-carbaldehyde
PubChem CID 79911
IUPAC Name 3-methylthiophene-2-carbaldehyde
SMILES CC1=C(SC=C1)C=O

Specifications

Boiling Point 113°C
Quantity 25 g
Formula Weight 126.17
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylthiophene-2-carboxaldehyde
EINECSNumber (5)-0074
TSCA No