missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)benzoic acid, 97%

Catalog No. AAH2720014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2720014 25 g
AAH2720003 1 g
AAH2720006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2720014 Supplier Thermo Scientific Chemicals Supplier No. H2720014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 825-99-0 | C8H8O2S | 168.21 g/mol

It is used in the de novo design of small molecule inhibitors targeting the LEDGF/p75-HIV integrase interaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the de novo design of small molecule inhibitors targeting the LEDGF/p75-HIV integrase interaction.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 825-99-0
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00156993
InChI Key PZGADOOBMVLBJE-UHFFFAOYSA-N
Synonym 3-methylthio benzoic acid, 3-methylsulfanyl benzoic acid, benzoic acid, 3-methylthio, benzoic acid, m-methylthio, 3-mercaptobenzoic acid s-methyl ether, 3-methylsulfanyl-benzoic acid, 3-carboxythioanisole, pubchem10679
PubChem CID 220329
IUPAC Name 3-methylsulfanylbenzoic acid
SMILES CSC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 126°C to 132°C
Quantity 25 g
Formula Weight 168.21
Percent Purity 97%
Chemical Name or Material 3-(Methylthio)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only