missing translation for 'onlineSavingsMsg'
Learn More

(3-Methylsulfonylaminomethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH5270503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5270503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5270503 Supplier Thermo Scientific Chemicals Supplier No. H5270503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

(3-Methylsulfonylaminomethyl)benzeneboronic acid, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Methylsulfonylaminomethyl)benzeneboronic acid, is used as a pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions

Chemical Identifiers

CAS 850568-39-7
Molecular Formula C8H12BNO4S
Molecular Weight (g/mol) 229.057
MDL Number MFCD06659817
InChI Key FJSWJJOLFPIDER-UHFFFAOYSA-N
Synonym 3-methylsulfonamidomethyl phenyl boronic acid, 3-methylsulphonylamino methyl benzeneboronic acid, 3-methanesulfonylaminomethylphenyl boronic acid, 3-methanesulfonylaminomethyl phenylboronic acid, 3-methanesulfonylaminomethyl benzeneboronic acid, 3-methanesulfonamidomethyl phenylboronic acid, 3-methylsulfonylaminomethyl benzeneboronic acid, 3-methylsulfonamidomethyl phenylboronic acid, 3-methylsulfonamidomethyl phenyl boronicacid, 3-methanesulfonamidomethyl phenyl boronic acid
PubChem CID 22309453
IUPAC Name [3-(methanesulfonamidomethyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)CNS(=O)(=O)C)(O)O

Specifications

Melting Point 246°C to 248°C
Quantity 1 g
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 229.06
Percent Purity 98%
Chemical Name or Material (3-Methylsulfonylaminomethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only