missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpyridine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. M11415G
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11415G 5 g
M1141100MG 100 mg
M11411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M11415G Supplier TCI America Supplier No. M11415G

Chemical Identifiers

CAS 4021-07-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00191553
InChI Key LMHIBYREWJHKNZ-UHFFFAOYSA-N
Synonym 3-methylpicolinic acid, 3-methyl-2-pyridinecarboxylic acid, 3-methyl-2-picolinic acid, 2-pyridinecarboxylic acid, 3-methyl, 3-methyl-a-picolinic acid, 3-methyl-alpha-picolinic acid, 3-methylpyridine-2-carboxylicacid, methylpyridincarbonsaure, pubchem14396, pubchem17659
PubChem CID 268757
IUPAC Name 3-methylpyridine-2-carboxylic acid
SMILES CC1=C(N=CC=C1)C(=O)O

Specifications

Melting Point 119°C
Color White
Quantity 5 g
Formula Weight 137.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylpyridine-2-carboxylic Acid